Bump cap, CAP 2000
mørkeblå
Katalog # 111-1322
Leverandør: CENTURION SAFETY PRODUCTS
Gå til:
Product Details & Documents
This bump cap protects against minor bumps and scrapes to the head. Ideal applications include automotive industries, transportation, shipping and track maintenance.
- Inner shell made of lightweight ABS, cap made of cotton and Lycra®
- Large crown ventilation holes with four mesh grills: Perfect for usage at higher temperatures
- Fabric covered EVA foam pad for improved wearing comfort
- Adjustable via velcro strap with tuck-in strap eyelet
For head sizes 54 to 59 cm.
Weight: 180 g
Certifications: EN 812 (−30 °C)
Specifikationer
- Farve:mørkeblå