Clamps, universal, slewable
0 - 80 mm
Katalog # 241-0349
Leverandør: USBECK Laborgeräte
Gå til:
Product Details & Documents
Universal clamp made of powder-coated zinc die-cast.With cork lined jaws.
- Capacity 0 to 80 mm
- Slewing range 180°
Specifikationer
- Spændvidde:0 - 80 mm