Shelving systems, wire, Super Erecta®
Om denne vare
The open wire design of these heavy-gauge shelves minimizes dust accumulation and allows a free circulation of air, greater visibility of stored items and greater light penetration.
- Fast, secure assembly: shelf is assembled in minutes without the use of any special tools
- Versatile construction: by using various accessories, hundreds of shelving configurations become possible
- Accessibility: shelves can be loaded/unloaded easily from all sides; this open construction allows use of maximum storage space of cube
- Adjustability: shelves can be adjusted 25 mm intervals along the entire length of the post
- Adjustable feet: bolt levelers compensate for surface irregularities
Brite: economical, chromate finish for dry, low humidity environments. Metro's Brite finish provides the look of chrome and includes a protective lacquer coat.
Chrome: the 'real' nickel-chrome finish for dry, low humidity environments. Metro's durable chrome finish includes a protective lacquer coat.
Type 304 polyshed stainless steel: addresses the most aggressive applications and environments. All-stainless solid and wire options exist for high temperature automated cart wash and autoclave applications.
Metroseal 3 with Microban: corrosion resistant finish for wet or high humidity environments with a 12 year warranty against rust and corrosion. Microban antimicrobial is built into the finish to keep the product 'cleaner between cleanings'.
: Plastic sleeves and wedges are included with each shelf.
72 tilgængelige muligheder nedenfor
Product Details & Documents
The open wire design of these heavy-gauge shelves minimizes dust accumulation and allows a free circulation of air, greater visibility of stored items and greater light penetration.
- Fast, secure assembly: shelf is assembled in minutes without the use of any special tools
- Versatile construction: by using various accessories, hundreds of shelving configurations become possible
- Accessibility: shelves can be loaded/unloaded easily from all sides; this open construction allows use of maximum storage space of cube
- Adjustability: shelves can be adjusted 25 mm intervals along the entire length of the post
- Adjustable feet: bolt levelers compensate for surface irregularities
Brite: economical, chromate finish for dry, low humidity environments. Metro's Brite finish provides the look of chrome and includes a protective lacquer coat.
Chrome: the 'real' nickel-chrome finish for dry, low humidity environments. Metro's durable chrome finish includes a protective lacquer coat.
Type 304 polyshed stainless steel: addresses the most aggressive applications and environments. All-stainless solid and wire options exist for high temperature automated cart wash and autoclave applications.
Metroseal 3 with Microban: corrosion resistant finish for wet or high humidity environments with a 12 year warranty against rust and corrosion. Microban antimicrobial is built into the finish to keep the product 'cleaner between cleanings'.
: Plastic sleeves and wedges are included with each shelf.
Product Family Options
Metroseal 3 with Microban
Produktinformation
- MaterialeL×BVægtTilgængelighedPrice
- METO1460NK3Super Erecta® shelving system: wire shelf, 1460NK3Super Erecta® shelving system: wire shelf, 1460NK3Metrosæl 31524×355 mm6,3 kgLog ind for tilgængelighed1 * 1 Items1.400,00 krSpecifikationer:
More
Specifications
Cat. No.METO1460NK3 - METO1824NK3Super Erecta® shelving system: wire shelf, 1824NK3Super Erecta® shelving system: wire shelf, 1824NK3Metrosæl 3610×457 mm3,2 kgLog ind for tilgængelighed1 * 1 Items1.150,00 krSpecifikationer:
More
Specifications
Cat. No.METO1824NK3 - METO1830NK3Super Erecta® shelving system: wire shelf, 1830NKSuper Erecta® shelving system: wire shelf, 1830NKMetrosæl 3760×457 mm3,6 kgLog ind for tilgængelighed1 * 1 Items1.210,00 krSpecifikationer:
More
Specifications
Cat. No.METO1830NK3 - METO1836NK3Super Erecta® shelving system: wire shelf, 1836NK3Super Erecta® shelving system: wire shelf, 1836NK3Metrosæl 3914×457 mm4,3 kgLog ind for tilgængelighed1 * 1 Items1.210,00 krSpecifikationer:
More
Specifications
Cat. No.METO1836NK3 - METO1842NK3Super Erecta® shelving system: wire shelf, 1842NK3Super Erecta® shelving system: wire shelf, 1842NK3Metrosæl 31066×457 mm5,0 kgLog ind for tilgængelighed1 * 1 Items1.440,00 krSpecifikationer:
More
Specifications
Cat. No.METO1842NK3 - METO1848NK3Super Erecta® shelving system: wire shelf, 1848NK3Super Erecta® shelving system: wire shelf, 1848NK3Metrosæl 31219×457 mm5,4 kgLog ind for tilgængelighed1 * 1 Items1.440,00 krSpecifikationer:
More
Specifications
Cat. No.METO1848NK3 - METO1854NK3Super Erecta® shelving system: wire shelf, 1854NK3Super Erecta® shelving system: wire shelf, 1854NK3Metrosæl 31370×457 mm6,6 kgLog ind for tilgængelighed1 * 1 Items1.700,00 krSpecifikationer:
More
Specifications
Cat. No.METO1854NK3 - METO1860NK3Super Erecta® shelving system: wire shelf, 1860NK3Super Erecta® shelving system: wire shelf, 1860NK3Metrosæl 31524×457 mm7,7 kgLog ind for tilgængelighed1 * 1 Items1.700,00 krSpecifikationer:
More
Specifications
Cat. No.METO1860NK3 - METO2442NK3Super Erecta® shelving system: wire shelf, 2442NK3Super Erecta® shelving system: wire shelf, 2442NK3Metrosæl 31066×610 mm6,8 kgLog ind for tilgængelighed1 * 1 Items1.780,00 krSpecifikationer:
More
Specifications
Cat. No.METO2442NK3 - METO2460NK3Super Erecta® shelving system: wire shelf, 2460NK3Super Erecta® shelving system: wire shelf, 2460NK3Metrosæl 31524×610 mm9,5 kgLog ind for tilgængelighed1 * 1 Items2.100,00 krSpecifikationer:
More
Specifications
Cat. No.METO2460NK3 - METO2472NK3Super Erecta® shelving system: wire shelf, 2472NK3Super Erecta® shelving system: wire shelf, 2472NK3Metrosæl 31829×610 mm11,8 kgLog ind for tilgængelighed1 * 1 Items2.480,00 krSpecifikationer:
More
Specifications
Cat. No.METO2472NK3
Chrome
Produktinformation
- MaterialeL×BVægtTilgængelighedPrice
- METO1424NCSuper Erecta® shelving system: wire shelf, 1424NCSuper Erecta® shelving system: wire shelf, 1424NCForkromede610×355 mm2,7 kgLog ind for tilgængelighed1 * 1 Items896,00 krSpecifikationer:
More
Specifications
Cat. No.METO1424NC - METO1430NCSuper Erecta® shelving system: wire shelf, 1430NCSuper Erecta® shelving system: wire shelf, 1430NCForkromede760×355 mm3,2 kgLog ind for tilgængelighed1 * 1 Items936,00 krSpecifikationer:
More
Specifications
Cat. No.METO1430NC - METO1436NCSuper Erecta® shelving system: wire shelf, 1436NCSuper Erecta® shelving system: wire shelf, 1436NCForkromede914×355 mm3,6 kgLog ind for tilgængelighed1 * 1 Items936,00 krSpecifikationer:
More
Specifications
Cat. No.METO1436NC - METO1442NCSuper Erecta® shelving system: wire shelf, 1442NCSuper Erecta® shelving system: wire shelf, 1442NCForkromede1066×355 mm4,3 kgLog ind for tilgængelighed1 * 1 Items1.080,00 krSpecifikationer:
More
Specifications
Cat. No.METO1442NC - METO1448NCSuper Erecta® shelving system: wire shelf, 1448NCSuper Erecta® shelving system: wire shelf, 1448NCForkromede1219×355 mm4,7 kgLog ind for tilgængelighed1 * 1 Items1.080,00 krSpecifikationer:
More
Specifications
Cat. No.METO1448NC - METO1460NCSuper Erecta® shelving system: wire shelf, 1460NCSuper Erecta® shelving system: wire shelf, 1460NCForkromede1524×355 mm6,3 kgLog ind for tilgængelighed1 * 1 Items1.270,00 krSpecifikationer:
More
Specifications
Cat. No.METO1460NC - METO1472NCSuper Erecta® shelving system: wire shelf, 1472NCSuper Erecta® shelving system: wire shelf, 1472NCForkromede1829×355 mm7,7 kgLog ind for tilgængelighed1 * 1 Items1.430,00 krSpecifikationer:
More
Specifications
Cat. No.METO1472NC - METO1818NCSuper Erecta® shelving system: wire shelf, 1818NCSuper Erecta® shelving system: wire shelf, 1818NCForkromede457×457 mm2,7 kgLog ind for tilgængelighed1 * 1 Items976,00 krSpecifikationer:
More
Specifications
Cat. No.METO1818NC - METO1824NCSuper Erecta® shelving system: wire shelf, 1824NCSuper Erecta® shelving system: wire shelf, 1824NCForkromede610×457 mm3,2 kgLog ind for tilgængelighed1 * 1 Items976,00 krSpecifikationer:
More
Specifications
Cat. No.METO1824NC - METO1830NCSuper Erecta® shelving system: wire shelf, 1830NCSuper Erecta® shelving system: wire shelf, 1830NCForkromede760×457 mm3,6 kgLog ind for tilgængelighed1 * 1 Items990,00 krSpecifikationer:
More
Specifications
Cat. No.METO1830NC - 139-0278Super Erecta® shelving system: wire shelf, 1836NCSuper Erecta® shelving system: wire shelf, 1836NCForkromede914×457 mm4,3 kgLog ind for tilgængelighed1 * 1 Items990,00 krSpecifikationer:
More
Specifications
Cat. No.139-0278 - 139-0279Super Erecta® shelving system: wire shelf, 1842NCSuper Erecta® shelving system: wire shelf, 1842NCForkromede1066×457 mm5,0 kgLog ind for tilgængelighed1 * 1 Items1.200,00 krSpecifikationer:
More
Specifications
Cat. No.139-0279 - 139-0280Super Erecta® shelving system: wire shelf, 1848NCSuper Erecta® shelving system: wire shelf, 1848NCForkromede1219×457 mm5,4Log ind for tilgængelighed1 * 1 Items1.200,00 krSpecifikationer:
More
Specifications
Cat. No.139-0280 - 139-0281Super Erecta® shelving system: wire shelf, 1854NCSuper Erecta® shelving system: wire shelf, 1854NCForkromede1370×457 mm6,6 kgLog ind for tilgængelighed1 * 1 Items1.390,00 krSpecifikationer:
More
Specifications
Cat. No.139-0281 - 139-0282Super Erecta® shelving system: wire shelf, 1860NCSuper Erecta® shelving system: wire shelf, 1860NCForkromede1524×457 mm7,7 kgLog ind for tilgængelighed1 * 1 Items1.390,00 krSpecifikationer:
More
Specifications
Cat. No.139-0282 - 139-0283Super Erecta® shelving system: wire shelf, 1872NCSuper Erecta® shelving system: wire shelf, 1872NCForkromede1829×457 mm9,1 kgLog ind for tilgængelighed1 * 1 Items1.690,00 krSpecifikationer:
More
Specifications
Cat. No.139-0283 - METO2124NCSuper Erecta® shelving system: wire shelf, 2124NCSuper Erecta® shelving system: wire shelf, 2124NCForkromede610×530 mm3,6 kgLog ind for tilgængelighed1 * 1 Items1.150,00 krSpecifikationer:
More
Specifications
Cat. No.METO2124NC - METO2130NCSuper Erecta® shelving system: wire shelf, 2130NCSuper Erecta® shelving system: wire shelf, 2130NCForkromede760×530 mm4,1 kgLog ind for tilgængelighed1 * 1 Items1.190,00 krSpecifikationer:
More
Specifications
Cat. No.METO2130NC - 139-0284Super Erecta® shelving system: wire shelf, 2136NCSuper Erecta® shelving system: wire shelf, 2136NCForkromede914×530 mm5,0 kgLog ind for tilgængelighed1 * 1 Items1.190,00 krSpecifikationer:
More
Specifications
Cat. No.139-0284 - 139-0285Super Erecta® shelving system: wire shelf, 2142NCSuper Erecta® shelving system: wire shelf, 2142NCForkromede1066×530 mm5,4 kgLog ind for tilgængelighed1 * 1 Items1.380,00 krSpecifikationer:
More
Specifications
Cat. No.139-0285 - 139-0286Super Erecta® shelving system: wire shelf, 2148NCSuper Erecta® shelving system: wire shelf, 2148NCForkromede1219×530 mm6,4Log ind for tilgængelighed1 * 1 Items1.380,00 krSpecifikationer:
More
Specifications
Cat. No.139-0286 - 139-0287Super Erecta® shelving system: wire shelf, 2154NCSuper Erecta® shelving system: wire shelf, 2154NCForkromede1370×530 mm7,3 kgLog ind for tilgængelighed1 * 1 Items1.640,00 krSpecifikationer:
More
Specifications
Cat. No.139-0287 - 139-0288Super Erecta® shelving system: wire shelf, 2160NCSuper Erecta® shelving system: wire shelf, 2160NCForkromede1524×530 mm8,2 kgLog ind for tilgængelighed1 * 1 Items1.640,00 krSpecifikationer:
More
Specifications
Cat. No.139-0288 - 139-0289Super Erecta® shelving system: wire shelf, 2172NCSuper Erecta® shelving system: wire shelf, 2172NCForkromede1829×530 mm10,9 kgLog ind for tilgængelighed1 * 1 Items1.950,00 krSpecifikationer:
More
Specifications
Cat. No.139-0289 - METO2424NCSuper Erecta® shelving system: wire shelf, 2424NCSuper Erecta® shelving system: wire shelf, 2424NCForkromede610×610 mm4,1 kgLog ind for tilgængelighed1 * 1 Items1.220,00 krSpecifikationer:
More
Specifications
Cat. No.METO2424NC - 139-0290Super Erecta® shelving system: wire shelf, 2436NCSuper Erecta® shelving system: wire shelf, 2436NCForkromede914×610 mm5,9 kgLog ind for tilgængelighed1 * 1 Items1.280,00 krSpecifikationer:
More
Specifications
Cat. No.139-0290 - 139-0291Super Erecta® shelving system: wire shelf, 2442NCSuper Erecta® shelving system: wire shelf, 2442NCForkromede1066×610 mm6,8 kgLog ind for tilgængelighed1 * 1 Items1.480,00 krSpecifikationer:
More
Specifications
Cat. No.139-0291 - 139-0292Super Erecta® shelving system: wire shelf, 2448NCSuper Erecta® shelving system: wire shelf, 2448NCForkromede1219×610 mm7,3 kgLog ind for tilgængelighed1 * 1 Items1.480,00 krSpecifikationer:
More
Specifications
Cat. No.139-0292 - 139-0293Super Erecta® shelving system: wire shelf, 2454NCSuper Erecta® shelving system: wire shelf, 2454NCForkromede1370×610 mm8,6 kgLog ind for tilgængelighed1 * 1 Items1.760,00 krSpecifikationer:
More
Specifications
Cat. No.139-0293 - 139-0294Super Erecta® shelving system: wire shelf, 2460NCSuper Erecta® shelving system: wire shelf, 2460NCForkromede1524×610 mm9,5 kgLog ind for tilgængelighed1 * 1 Items1.760,00 krSpecifikationer:
More
Specifications
Cat. No.139-0294 - 139-0295Super Erecta® shelving system: wire shelf, 2472NCSuper Erecta® shelving system: wire shelf, 2472NCForkromede1829×610 mm11,8 kgLog ind for tilgængelighed1 * 1 Items2.100,00 krSpecifikationer:
More
Specifications
Cat. No.139-0295 - METO3060NCSuper Erecta® shelving system: wire shelf, 3060NCSuper Erecta® shelving system: wire shelf, 3060NCForkromede1524×760 mm11,8 kgLog ind for tilgængelighed1 * 1 Items2.370,00 krSpecifikationer:
More
Specifications
Cat. No.METO3060NC - METO3072NCSuper Erecta® shelving system: wire shelf, 3072NCSuper Erecta® shelving system: wire shelf, 3072NCForkromede1829×760 mm14,0 kgLog ind for tilgængelighed1 * 1 Items2.830,00 krSpecifikationer:
More
Specifications
Cat. No.METO3072NC - METO3648NCSuper Erecta® shelving system: wire shelf, 3648NCSuper Erecta® shelving system: wire shelf, 3648NCForkromede1219×910 mm10,4 kgLog ind for tilgængelighed1 * 1 Items2.490,00 krSpecifikationer:
More
Specifications
Cat. No.METO3648NC
Stainless steel
Produktinformation
- MaterialeL×BVægtTilgængelighedPrice
- METO1424NSSuper Erecta® shelving system: wire shelf, 1424NSSuper Erecta® shelving system: wire shelf, 1424NSRustfrit stål610×355 mm2,7 kgLog ind for tilgængelighed1 * 1 Items2.780,00 krSpecifikationer:
More
Specifications
Cat. No.METO1424NS - METO1430NSSuper Erecta® shelving system: wire shelf, 1430NSSuper Erecta® shelving system: wire shelf, 1430NSRustfrit stål760×355 mm3,2 kgLog ind for tilgængelighed1 * 1 Items2.900,00 krSpecifikationer:
More
Specifications
Cat. No.METO1430NS - METO1436NSSuper Erecta® shelving system: wire shelf, 1436NSSuper Erecta® shelving system: wire shelf, 1436NSRustfrit stål914×355 mm3,6 kgLog ind for tilgængelighed1 * 1 Items2.700,00 krSpecifikationer:
More
Specifications
Cat. No.METO1436NS - METO1460NSSuper Erecta® shelving system: wire shelf, 1460NSSuper Erecta® shelving system: wire shelf, 1460NSRustfrit stål1524×355 mm6,3 kgLog ind for tilgængelighed1 * 1 Items4.470,00 krSpecifikationer:
More
Specifications
Cat. No.METO1460NS - METO1472NSSuper Erecta® shelving system: wire shelf, 1472NSSuper Erecta® shelving system: wire shelf, 1472NSRustfrit stål1829×355 mm7,7 kgLog ind for tilgængelighed1 * 1 Items5.030,00 krSpecifikationer:
More
Specifications
Cat. No.METO1472NS - METO1824NSSuper Erecta® shelving system: wire shelf, 1824NSSuper Erecta® shelving system: wire shelf, 1824NSRustfrit stål610×457 mm3,2 kgLog ind for tilgængelighed1 * 1 Items3.510,00 krSpecifikationer:
More
Specifications
Cat. No.METO1824NS - 139-0296Super Erecta® shelving system: wire shelf, 1836NSSuper Erecta® shelving system: wire shelf, 1836NSRustfrit stål914×457 mm4,3 kgLog ind for tilgængelighed1 * 1 Items3.550,00 krSpecifikationer:
More
Specifications
Cat. No.139-0296 - 139-0297Super Erecta® shelving system: wire shelf, 1842NSSuper Erecta® shelving system: wire shelf, 1842NSRustfrit stål1066×457 mm5,0 kgLog ind for tilgængelighed1 * 1 Items4.200,00 krSpecifikationer:
More
Specifications
Cat. No.139-0297 - 139-0298Super Erecta® shelving system: wire shelf, 1848NSSuper Erecta® shelving system: wire shelf, 1848NSRustfrit stål1219×457 mm5,4 kgLog ind for tilgængelighed1 * 1 Items4.200,00 krSpecifikationer:
More
Specifications
Cat. No.139-0298 - 139-0308Super Erecta® shelving system: wire shelf, 2148NSSuper Erecta® shelving system: wire shelf, 2148NSRustfrit stål1219×457 mm6,4 kgLog ind for tilgængelighed1 * 1 Items4.720,00 krSpecifikationer:
More
Specifications
Cat. No.139-0308 - 139-0299Super Erecta® shelving system: wire shelf, 1854NSSuper Erecta® shelving system: wire shelf, 1854NSRustfrit stål1370×457 mm6,6 kgLog ind for tilgængelighed1 * 1 Items5.330,00 krSpecifikationer:
More
Specifications
Cat. No.139-0299 - 139-0304Super Erecta® shelving system: wire shelf, 1860NSSuper Erecta® shelving system: wire shelf, 1860NSRustfrit stål1524×457 mm7,7 kgLog ind for tilgængelighed1 * 1 Items5.330,00 krSpecifikationer:
More
Specifications
Cat. No.139-0304 - 139-0305Super Erecta® shelving system: wire shelf, 1872NSSuper Erecta® shelving system: wire shelf, 1872NSRustfrit stål1829×457 mm9,1 kgLog ind for tilgængelighed1 * 1 Items6.330,00 krSpecifikationer:
More
Specifications
Cat. No.139-0305 - METO2124NSSuper Erecta® shelving system: wire shelf, 2124NSSuper Erecta® shelving system: wire shelf, 2124NSRustfrit stål610×530 mm3,6 kgLog ind for tilgængelighed1 * 1 Items4.070,00 krSpecifikationer:
More
Specifications
Cat. No.METO2124NS - METO2130NSSuper Erecta® shelving system: wire shelf, 2130NSSuper Erecta® shelving system: wire shelf, 2130NSRustfrit stål760×530 mm4,1 kgLog ind for tilgængelighed1 * 1 Items3.830,00 krSpecifikationer:
More
Specifications
Cat. No.METO2130NS - 139-0306Super Erecta® shelving system: wire shelf, 2136NSSuper Erecta® shelving system: wire shelf, 2136NSRustfrit stål914×530 mm5,0 kgLog ind for tilgængelighed1 * 1 Items4.110,00 krSpecifikationer:
More
Specifications
Cat. No.139-0306 - 139-0307Super Erecta® shelving system: wire shelf, 2142NSSuper Erecta® shelving system: wire shelf, 2142NSRustfrit stål1066×530 mm5,4 kgLog ind for tilgængelighed1 * 1 Items4.720,00 krSpecifikationer:
More
Specifications
Cat. No.139-0307 - 139-0309Super Erecta® shelving system: wire shelf, 2154NSSuper Erecta® shelving system: wire shelf, 2154NSRustfrit stål1370×530 mm7,3 kgLog ind for tilgængelighed1 * 1 Items6.260,00 krSpecifikationer:
More
Specifications
Cat. No.139-0309 - 139-0310Super Erecta® shelving system: wire shelf, 2160NSSuper Erecta® shelving system: wire shelf, 2160NSRustfrit stål1524×530 mm8,2 kgLog ind for tilgængelighed1 * 1 Items6.260,00 krSpecifikationer:
More
Specifications
Cat. No.139-0310 - 139-0311Super Erecta® shelving system: wire shelf, 2172NSSuper Erecta® shelving system: wire shelf, 2172NSRustfrit stål1829×530 mm10,9 kgLog ind for tilgængelighed1 * 1 Items7.400,00 krSpecifikationer:
More
Specifications
Cat. No.139-0311 - 139-0312Super Erecta® shelving system: wire shelf, 2436NSSuper Erecta® shelving system: wire shelf, 2436NSRustfrit stål914×610 mm5,9 kgLog ind for tilgængelighed1 * 1 Items4.370,00 krSpecifikationer:
More
Specifications
Cat. No.139-0312 - 139-0313Super Erecta® shelving system: wire shelf, 2442NSSuper Erecta® shelving system: wire shelf, 2442NSRustfrit stål1066×610 mm6,8 kgLog ind for tilgængelighed1 * 1 Items5.280,00 krSpecifikationer:
More
Specifications
Cat. No.139-0313 - 139-0314Super Erecta® shelving system: wire shelf, 2448NSSuper Erecta® shelving system: wire shelf, 2448NSRustfrit stål1219×610 mm7,3 kgLog ind for tilgængelighed1 * 1 Items5.280,00 krSpecifikationer:
More
Specifications
Cat. No.139-0314 - 139-0315Super Erecta® shelving system: wire shelf, 2454NSSuper Erecta® shelving system: wire shelf, 2454NSRustfrit stål1370×610 mm8,6 kgLog ind for tilgængelighed1 * 1 Items6.430,00 krSpecifikationer:
More
Specifications
Cat. No.139-0315 - 139-0316Super Erecta® shelving system: wire shelf, 2460NSSuper Erecta® shelving system: wire shelf, 2460NSRustfrit stål1524×610 mm9,5 kgLog ind for tilgængelighed1 * 1 Items6.430,00 krSpecifikationer:
More
Specifications
Cat. No.139-0316 - 139-0317Super Erecta® shelving system: wire shelf, 2472NSSuper Erecta® shelving system: wire shelf, 2472NSRustfrit stål1829×610 mm11,8 kgLog ind for tilgængelighed1 * 1 Items7.990,00 krSpecifikationer:
More
Specifications
Cat. No.139-0317 - METO3048NSSuper Erecta® shelving system: wire shelf, 3048NSSuper Erecta® shelving system: wire shelf, 3048NSRustfrit stål1219×760 mm9,5 kgLog ind for tilgængelighed1 * 1 Items6.970,00 krSpecifikationer:
More
Specifications
Cat. No.METO3048NS