ContactUsLinkComponent
Buffer, SSC solution 20X concentrate (saline-sodium citrate)
Buffer, SSC solution 20X concentrate (saline-sodium citrate)
Catalog # R020
Supplier:  G-Biosciences
CAS Number:  
Buffer, SSC solution 20X concentrate (saline-sodium citrate)
Catalog # R020
Supplier:  G-Biosciences
CAS Number:  
Specifications
  • Pakn:
    4 L
  • Beskrivelse:
    SSC buffer solution 20X concentrate (saline-sodium citrate)