Free fat butyrometer
Butyrometer for free fat, complete with screw cap, scale 0,002 g
Katalog # 614-0136
Leverandør: GERBER FUNKE
Gå til:
Product Details & Documents
For determining free fat content in milk and cream.
Specifikationer
- Beskrivelse:Butyrometer for free fat, complete with screw cap, scale 0,002 g