Precision butyrometer, without stopper
Leverandør: GERBER FUNKE
Gå til:
Om denne vare
For drinking vat milk, frosted rear scale wall, error tolerance 0,025%.
4 tilgængelige muligheder nedenfor
Product Details & Documents
For drinking vat milk, frosted rear scale wall, error tolerance 0,025%.
Product Family Options
Produktinformation
- TilgængelighedPrice
- 614-0088Butyrometer, precision, 0 - 4 % : 0,05
- GERB3150-CButyrometer, precision, 0 - 4 % : 0,05, with conformity certificateButyrometer, precision, 0 - 4 % : 0,05, with conformity certificateLog ind for tilgængelighed1 * 1 Items249,00 kr
- GERB3150-C-OButyrometer, precision, 0 - 4 % : 0,05, with conformity certificate, without stopperButyrometer, precision, 0 - 4 % : 0,05, with conformity certificate, without stopperLog ind for tilgængelighed1 * 1 Items243,00 kr
- GERB3150-OButyrometer, precision, 0 - 4 % : 0,05, without stopperButyrometer, precision, 0 - 4 % : 0,05, without stopperLog ind for tilgængelighed1 * 10 Items936,00 kr
Accessories
Produktinformation
- Katalog #TilgængelighedPrice
- Stopper, rubber, conical, for all measuring method butyrometersStopper, rubber, conical, for all measuring method butyrometers614-2028Log ind for tilgængelighed1 * 100 Items - 360,00 kr