Orders LinkContactUsLinkComponent
VWR®RapidBlock™, Blocking solution, 10x
VWR®RapidBlock™, Blocking solution, 10x
RapidBlock™ solution, 10X, 100 mL
Katalog # M325-100ML
Leverandør:   VWR Chemicals
VWR®RapidBlock™, Blocking solution, 10x
Katalog # M325-100ML
Leverandør:  VWR Chemicals
CAS nummer:  
VWR®RapidBlock™, Blocking solution, 10x
VWR®RapidBlock™, Blocking solution, 10x
RapidBlock™ solution, 10X, 100 mL
CAS nummer:  
Price
2.440,00 kr
Log ind for tilgængelighed
1 * 100 mL
Product Details & Documents
RapidBlock™ solution reduces blocking time to five minutes for Western and dot blotting procedures on PVDF and nitrocellulose membranes. This protein-free formulation minimises cross-reactivity and non specific antibody binding, generating blots with low background and enhanced signal-to-noise ratio.
  • Only 5 minutes for PVDF or nitrocellulose blocking
  • Reduce background and cross-reactivity
  • Enhance chemiluminescent signals
  • Protein-free formulation
Specifikationer
Specifikationer
  • Pakn:
    100 mL
  • Beskrivelse:
    RapidBlock™ solution, 10X