Orders LinkContactUsLinkComponent
VWR®, Destaining bags
VWR®, Destaining bags
Destaining bags, 25
Katalog # E732-25
Leverandør:   VWR Chemicals
VWR®, Destaining bags
Katalog # E732-25
Leverandør:  VWR Chemicals
CAS nummer:  
VWR®, Destaining bags
VWR®, Destaining bags
Destaining bags, 25
CAS nummer:  
Price
1.030,00 kr
Log ind for tilgængelighed
1 * 25 Items
Product Details & Documents
Destaining bags are designed to safely remove ethidium bromide from solution during overnight treatment. Each bag extracts up to 5 mg of ethidium bromide from solution, thus minimising the exposure of research personnel to this toxic material. The special absorbent mixture retains dye molecules in a convenient bag for removal and incineration.
  • Removes up to 5 mg ethidium bromide per bag
  • Minimises exposure to toxic materials
  • Easiest ethidium bromide removal available
Specifikationer
Specifikationer
  • Pakn:
    25
  • Beskrivelse:
    Destaining bags