Pesticid referencestandard, Malathion (1,2-Bis(ethoxycarbonyl)ethyl O,O-dimethyl phosphorodithioate) 100 µg/ml in n-nonane
Catalog # CAMBULM-8122-1.2
Supplier: CAMBRIDGE ISOTOPE LABS
CAS Number:
Specifications
- Pakning:Glass bottle
- Pakn:1,2 mL
- CAS-nr:121-75-5
- Beskrivelse:Malathion (1,2-Bis(ethoxycarbonyl)ethyl O,O-dimethyl phosphorodithioate) 100 µg/ml in n-nonane
- Formel:C₁₀H₁₉O₆PS₂
- Beskrivelse:Malathion (1,2-Bis(ethoxycarbonyl)ethyl O,O-dimethyl phosphorodithioate)