Forceps, SP Bel-Art
Buet, afrundet, 127 mm
Katalog # BELAF379200000
Leverandør: Bel-Art Products, a Part of SP
Gå til:
Product Details & Documents
PP, white.
- Strong and durable
- Serrated tips ensure a firm grasp
- Autoclavable up to 121 °C
Specifikationer
- Model:Buet, afrundet
- Længde:127 mm