Forceps, SP Bel-Art
Lige, spids, 98 mm
Catalog # BELAH379370000
Supplier: Bel-Art Products, a Part of SP
Jump to:
Product Details & Documents
Stainless steel, Teflon® FEP coated (partly of fully). The brown Teflon® FEP coating on these stainless steel forceps will not scratch or mar delicate surfaces, while its anti-stick qualities prevent material build-up.
- Coating complies with FDA regulations for direct contact with food
- All forceps are 98 mm (3⅞") long with medium points and serrated tips
Specifications
- Model:Lige, spids
- Længde:98 mm
- Seq:03