Goggles, uvex 9405
Anti-UV, anti-fog, Klar celluloseacetat, gennemsigtig grå
Catalog # 111-2613
Supplier: UVEX
Jump to:
Product Details & Documents
The large goggles are suitable to use with prescription eye wear and filtering half masks.
- Soft face and nose padding ensures comfortable fit
- Frosted upper part protects against light reflection
- Easy headband adjustment
Certifications: EN 166, EN 170
Specifications
- Model:Anti-UV, anti-fog
- Farve:gennemsigtig grå
- Linser:Klar celluloseacetat
Accessories
Product Information
- Catalog #BeskrivelseAvailabilityPrice