Orders LinkContactUsLinkComponent
Accessories for columns INdEX™
Leverandør:   Cytiva
Accessories for columns INdEX™
Leverandør:  Cytiva
Certifikater
Accessories for columns INdEX™
Accessories for columns INdEX™
5 tilgængelige muligheder nedenfor
Product Family Options
Produktinformation
  • Tilgængelighed
    Price
  • 18-1012-69
    Connector, 25 mm TC, I.Ø 22 mm, 51 mm TC, for INdEX™ 70, INdEX™ 100, INdEX™ 140 and INdEX™ 200 columns
    Connector, 25 mm TC, I.Ø 22 mm, 51 mm TC, for INdEX™ 70, INdEX™ 100, INdEX™ 140 and INdEX™ 200 columns
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - kr2,920.00
    kr2,920.00
    1 * 1 Items
       kr2,920.00
  • 18-1031-09
    Connector, 25 mm TC-M6 threaded, for INdEX™ 70 and INdEX™ 100 columns
    Connector, 25 mm TC-M6 threaded, for INdEX™ 70 and INdEX™ 100 columns
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 2 Items
    1 * 2 Items - kr778.00
    kr778.00
    1 * 2 Items
       kr778.00
  • 18-0019-27
    Gasket, EPDM, 25 mm, I.Ø 6 mm, for INdEX™ 70, INdEX™ 100, INdEX™ 140 and INdEX™ 200 columns
    Gasket, EPDM, 25 mm, I.Ø 6 mm, for INdEX™ 70, INdEX™ 100, INdEX™ 140 and INdEX™ 200 columns
    Discontinued
  • 18-0251-77
    Net, 10 μm, end-piece, PA, for INdEX™ 100 column
    Net, 10 μm, end-piece, PA, for INdEX™ 100 column
    Discontinued
  • 18-9252-01
    Net, 23 μm, end-piece, PP, for INdEX™ 100 columns
    Net, 23 μm, end-piece, PP, for INdEX™ 100 columns
    Discontinued
  • 18-1104-29
    T-junction, for INdEX™ 70, INdEX™ 100, INdEX™ 140 and INdEX™ 200 columns
    T-junction, for INdEX™ 70, INdEX™ 100, INdEX™ 140 and INdEX™ 200 columns
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - kr4,450.00
    kr4,450.00
    1 * 1 Items
       kr4,450.00
  • 18-0005-45
    Tubing with sanitary fitting, PVC, 25 mm TC, (I.Ø×L) 6×1500 mm, for INdEX™ 70, INdEX™ 100, INdEX™ 140 and INdEX™ 200 columns
    Tubing with sanitary fitting, PVC, 25 mm TC, (I.Ø×L) 6×1500 mm, for INdEX™ 70, INdEX™ 100, INdEX™ 140 and INdEX™ 200 columns
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - kr3,400.00
    kr3,400.00
    1 * 1 Items
       kr3,400.00
  • 18-0005-47
    Tubing with sanitary fitting, PVC, 25 mm TC, (I.Ø×L) 6×2000 mm, for INdEX™ 70, INdEX™ 100, INdEX™ 140 and INdEX™ 200 columns
    Tubing with sanitary fitting, PVC, 25 mm TC, (I.Ø×L) 6×2000 mm, for INdEX™ 70, INdEX™ 100, INdEX™ 140 and INdEX™ 200 columns
    Discontinued
  • 18-0005-42
    Tubing with sanitary fitting, PVC, 25 mm TC, (I.Ø×L) 6×300 mm, for INdEX™ 70, INdEX™ 100, INdEX™ 140 and INdEX™ 200 columns
    Tubing with sanitary fitting, PVC, 25 mm TC, (I.Ø×L) 6×300 mm, for INdEX™ 70, INdEX™ 100, INdEX™ 140 and INdEX™ 200 columns
    Discontinued
  • 18-5758-01
    Valves, 4-port, 4-way, for INdEX™ 70, INdEX™ 100, INdEX™ 140 and INdEX™ 200 columns
    Valves, 4-port, 4-way, for INdEX™ 70, INdEX™ 100, INdEX™ 140 and INdEX™ 200 columns
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - kr14,380.00
    kr14,380.00
    1 * 1 Items
       kr14,380.00