Order Entry
Denmark
ContactUsLinkComponent
Dichlorvos-D6 (dimethyl-D6) (2,2-Dichlorvinyl dimethylphosphat-D6, DDVP-D6) (98% D)
Dichlorvos-D6 (dimethyl-D6) (2,2-Dichlorvinyl dimethylphosphat-D6, DDVP-D6) (98% D)
Catalog # CAMBDLM-2829-0.01
CAS Number:  
Dichlorvos-D6 (dimethyl-D6) (2,2-Dichlorvinyl dimethylphosphat-D6, DDVP-D6) (98% D)
Catalog # CAMBDLM-2829-0.01
CAS Number:  

Specifications

  • Pakning:
    Glass bottle for solids
  • Pakn:
    0,1 g
  • Seq:
    3020

Specifications

About this item

 :
 :226.93 g/mol
CAS Number:203645-53-8
Formula:C₄HCl₂D₆O₄P