Pesticid referencestandard, Carbaryl (1-Naphthyl methylcarbamat) 100 µg/ml i n-nonan
Catalog # CAMBULM-8096-1.2
Supplier: CAMBRIDGE ISOTOPE LABS
CAS Number:
Specifications
- Pakning:Glass bottle
- Pakn:1,2 mL
- CAS-nr:63-25-2
- Beskrivelse:Carbaryl (1-Naphthyl methylcarbamat) 100 µg/ml i n-nonan
- Formel:C₁₂H₁₁NO₂
- Beskrivelse:Carbaryl (1-Naphthyl methylcarbamate)