Pesticid referencestandard, Dichlorvos (2,2-Dichlorvinyl dimethylphosphat, DDVP) 100 µg/ml i n-nonan
CAMBULM-7217-1.2
:
- Pakning:Glass bottle
- Pakn:1,2 mL
- CAS-nr:62-73-7
- Beskrivelse:Dichlorvos (2,2-Dichlorvinyl dimethylphosphat, DDVP) 100 µg/ml i n-nonan
- Formel:C₄H₇Cl₂O₄P
- Beskrivelse:Dichlorvos (2,2-Dichlorovinyl dimethyl phosphate, DDVP)