Order Entry
Denmark
ContactUsLinkComponent
 
Pesticid referencestandard, Hexachlorphen 50 µg/ml i methanol
  CAMBULM-8009-1.2
 :  
Pesticid referencestandard, Hexachlorphen 50 µg/ml i methanol
  CAMBULM-8009-1.2
 :  

 

  • Pakning:
    Glass bottle
  • Pakn:
    1,2 mL
  • CAS-nr:
    70-30-4
  • Beskrivelse:
    Hexachlorphen 50 µg/ml i methanol
  • Formel:
    C₁₃H₆Cl₆O₂
  • Beskrivelse:
    Hexachlorophene