NovaTaq PCR master mix
71007-3
:
- Pakn:250 EU
- Beskrivelse:NovaTaq PCR Master Mix
The NovaTaq PCR Master Mix is a ready to use 2X concentrated mixture of NovaTaq DNA polymerase, ultra-pure deoxynucleotides, and reaction buffer without magnesium chloride. The master mix simplifies the assembly of PCR reactions and offers advantages of times savings, consistency, and minimal risk of contamination. Simply add the NovaTaq PCR master mix to an equal volume containing the required amount of magnesium chloride, DNA template, and primers, and the reaction is ready for thermal cycling. The final diluted reaction contains 2,5 units of NovaTaq DNA polymerase per 100 µl. Sufficient components are included for 200 standard 50 µl (or 100×100 µl) amplification reactions.