Orders LinkContactUsLinkComponent
Tris-Acetate-EDTA (TAE) buffer, 50X
Tris-Acetate-EDTA (TAE) buffer, 50X
Tris-Acetate-EDTA (TAE) buffer, 50X, Glass bottle, 1 L
Katalog # A1691.1000
Leverandør:   PanReac AppliChem
Tris-Acetate-EDTA (TAE) buffer, 50X
Katalog # A1691.1000
Leverandør:  PanReac AppliChem
CAS nummer:  
Tris-Acetate-EDTA (TAE) buffer, 50X
Tris-Acetate-EDTA (TAE) buffer, 50X, Glass bottle, 1 L
CAS nummer:  
Price
971,00 kr
Log ind for tilgængelighed
1 * 1 L
Product Details & Documents
50X concentrated solution of 2 M Tris, 1 M glacial acetic acid and 0,5 M EDTA.Na₂, pH 8,5 ±0,2.
Documents
Specifikationer
Specifikationer
  • Pakning:
    Glass bottle
  • Pakn:
    1 L
  • Beskrivelse:
    Tris-Acetate-EDTA (TAE) buffer, 50X