Reagent Kits til Protein og Fibre Analyser
Katalog # 1.12979.0001
Leverandør: Merck
CAS nummer:
Specifikationer
- Pakn:100 Tests
- Beskrivelse:Samlet kostfibre
- Analyser:100
Specifikationer
Om denne vare
Simple reagent kits, used in food analysis, that use colour chemistry to determine levels of protein and fibre. Please note these kits require the use of a photometer or spectrophotometer.