Enkeltkanalpipette, mekanisk, fast eller variabelt volumen, Finnpipette™ F2
Supplier: Thermo Fisher Scientific
Certificates
About this item
Finnpipette™ F2 offers superior robustness and durability even in the most demanding applications.
- Fully autoclavable - No need to dismantle before autoclaving
- Made of high-quality PVDF that withstands exposure to chemicals and UV light
- Advanced Volume Gearing mechanism (AVG) - self-supporing modular mechanism increases accuracy, precision and durability, with light force and smaller increments for volume adjustment
- Large, contrasted Ergovisio display with white numbers on black background for increased volume visibility and security
- Soft-touch tip ejection with very light force
- Superior ergonomics and very low force pipetting movement
- Super blow-out piston for volumes of 50 µl and below, which ensures the delivery of micro-size drops
* Micro
GLP-Kits supplied with three Finnpipette® F2 (GLP-Kit 1, 3, 4) or four Finnpipette® F2 (GLP-Kit 2), one stand for six pipettes, three racks of 96 Finntip Flex tips and 15 disposable reagent reservoirs.
Ordering information: Supplied with one opening and calibration key, calibration report, three years warranty certificate (extended to five years with web registration) and user guide (English, French, German and Spanish).
GLP-Kits supplied with three Finnpipette™ F2 (GLP-Kit 1, 3, 4) or four Finnpipette™ F2 (GLP-Kit 2), one stand for six pipettes, three racks of 96 Finntip Flex tips and 15 disposable reagent reservoirs.
GLP-Kits supplied with three Finnpipette™ F2 (GLP-Kit 1, 3, 4) or four Finnpipette™ F2 (GLP-Kit 2), one stand for six pipettes, three racks of 96 Finntip Flex tips and 15 disposable reagent reservoirs.
Specifications
- Catalog No:
- 613-2708
- 613-2709
- 613-1480
- 613-1481
- 613-2710
- 613-2697
- 613-2714
- 613-2715
- 613-2716
- 613-2711
- 613-2712
- 613-2713
- 613-2696
- 613-2717
- 613-2698
- 613-2699
- 613-1486
- 613-2718
- 613-1487
- 613-2719
- 613-2704
- 613-2705
- 613-2706
- 613-2707
- 613-2700
- 613-2701
- 613-2702
- 613-2703
- Environmentally Preferable:
- Lav fremstillingspåvirkning
- Lav fremstillingspåvirkning
- Lav fremstillingspåvirkning
- Lav fremstillingspåvirkning
- Lav fremstillingspåvirkning
- Lav fremstillingspåvirkning
- Lav fremstillingspåvirkning
- Lav fremstillingspåvirkning
- Lav fremstillingspåvirkning
- Lav fremstillingspåvirkning
- Lav fremstillingspåvirkning
- Lav fremstillingspåvirkning
- Lav fremstillingspåvirkning
- Lav fremstillingspåvirkning
- Lav fremstillingspåvirkning
- Lav fremstillingspåvirkning
- Lav fremstillingspåvirkning
- Lav fremstillingspåvirkning
- Lav fremstillingspåvirkning
- Lav fremstillingspåvirkning
- Lav fremstillingspåvirkning
- Lav fremstillingspåvirkning
- Lav fremstillingspåvirkning
- Lav fremstillingspåvirkning
- Lav fremstillingspåvirkning
- Lav fremstillingspåvirkning
- Lav fremstillingspåvirkning
- Lav fremstillingspåvirkning
- Miljøcertificering:
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- IRECS
- ISO 14001 (Certificate Registration No. EMS 623400)
- IRECS
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- IRECS
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- IRECS
- IRECS
- IRECS
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- ISO 14001 (Certificate Registration No. EMS 623400)
- IRECS
- IRECS
Specifications
GLP Kits
Product Information
Frequently Bought Together