Staining jars acc. to Coplin, WHEATON®
Catalog # 720-0700
Supplier: DWK Life Sciences
Specifications
- Beskrivelse:Farvekar iht. Coplin, med glaslåg
- W×D×H:28×26×92 mm
- Environmentally Preferable:
- Pakn:1
Specifications
About this item
Staining jars, made of soda-lime glass, with extra tall lid.
It holds five single 75×25 mm and 75×100 mm slides vertically, or 10 slides back to back.