Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Denmark
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Lyskilder
UV-lamper
UV fraud detection lamp
UV fraud detection lamp
UV fraud detection lamp
Catalog # UVPA95-0164-02
Supplier:
UVP ULTRA VIOLET PRODUCTS
Expand image
UV fraud detection lamp
Catalog # UVPA95-0164-02
Supplier:
UVP ULTRA VIOLET PRODUCTS
Specifications
Til:
Dokument- og signaturverifikation
Beskrivelse:
UV-lampe SL-2M, BLB
Model:
SL-2M
Udgang:
4 W
Pakn:
1
Bølgelængde:
365 nm
Specifications
About this item
Product Details
Product Details