Orders LinkContactUsLinkComponent
Counting chambers
Counting chambers
Leverandør:   MARIENFELD
Counting chambers
Leverandør:  MARIENFELD
Certifikater
Counting chambers
Counting chambers
3 tilgængelige muligheder nedenfor
Product Details & Documents
These counting chambers are suitable for counting leucocytes and erythrocytes.
  • Double net ruling
  • Two counting grids with one division
  • Different types of design available
Certifications: IVD 98/79/EC.
Leveringsinformation: Supplied with two cover glasses of 0,4 mm thickness.
Documents
Product Family Options
Produktinformation
  • Model
    Kammerdybde
    Tilgængelighed
    Price
  • 630-1510
    Celletæller, mørke kanter
    Celletæller, mørke kanter
    Fuchs-Rosenthal
    0,2 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 439,00 kr
    439,00 kr
    1 * 1 Items
       439,00 kr
  • 630-1511
    Counting chamber, dark lined
    Counting chamber, dark lined
    Nageotte
    0,5 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 1.350,00 kr
    1.350,00 kr
    1 * 1 Items
       1.350,00 kr
  • 630-1509
    Counting chamber, bright lined
    Counting chamber, bright lined
    Imp. Neubauer
    0,1 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 880,00 kr
    880,00 kr
    1 * 1 Items
       880,00 kr