Orders LinkContactUsLinkComponent
Vials with insert, WHEATON®
Vials with insert, WHEATON®
Leverandør:   DWK Life Sciences
Vials with insert, WHEATON®
Leverandør:  DWK Life Sciences
Certifikater
Vials with insert, WHEATON®
Vials with insert, WHEATON®
2 tilgængelige muligheder nedenfor
Product Details & Documents
These vials made of PP uses either aluminum crimp seals or plastic snap cap closures, which eliminate the need for crimping and decapping tools, and provide a larger target area.
  • Available with snap/crimp or screw top finish
  • Unique cone-shaped bottom ensures maximum sample retrieval
Packaging: Packaged as 100 vials per shelf pack or 1000 vials per case.
Product Family Options
Produktinformation
  • Kapacitet
    Diameter×H
    Kasket inkluderet
    Låg type
    Tilgængelighed
    Price
  • WHEA225235
    Vials with insert
    Vials with insert
    0,75 ml
    12×32 mm
    No
    Snap/crimp
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1.000 Items
    1 * 1.000 Items - 3.020,00 kr
    3.020,00 kr
    1 * 1.000 Items
       3.020,00 kr
  • WHEAW225187
    Vials with insert
    Vials with insert
    0,50 ml
    12×32 mm
    No
    Snap/crimp
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1.000 Items
    1 * 1.000 Items - 3.010,00 kr
    3.010,00 kr
    1 * 1.000 Items
       3.010,00 kr