Accessories for empty columns C series
Leverandør: Cytiva
Gå til:
Om denne vare
15 tilgængelige muligheder nedenfor
Product Details & Documents
Documents
Cytiva CyDye Licensing Statements Cytiva General Licensing Statements
Additional Product Details
Product Family Options
Produktinformation
- TilgængelighedPrice
- 19-5006-01AC 10 flow adapter, for C-10 columns
- 19-5109-01AC 16 flow adapter, for C-16 columns
- 19-5207-01Flow adapters AC 26
- 19-5020-01Friction ring, C 10, silicone rubber
- 19-5008-01Net rings C 10, AC 10 (10 µm)
- 18-8761-01Net rings for columns, 16 mm I-Ø, 10 µm
- 19-0163-01O-rings for flow adapters and bottom end pieces
- 19-5043-01O-rings for flow adapters and bottom end pieces (AC 10)O-rings for flow adapters and bottom end pieces (AC 10)Log ind for tilgængelighed1 * 10 Items755,00 kr
- 19-0688-01O-rings for flow adapters and bottom end pieces, AK 26O-rings for flow adapters and bottom end pieces, AK 26Log ind for tilgængelighed1 * 10 Items432,00 kr
- 19-5007-01Packing reservoirs RC 10
- 19-5110-01Packing reservoirs RC 16
- 19-5208-01Packing reservoirs RC 26
- 19-5035-01Spare chromatographic tubes C 10/10
- 19-1767-01Support screen for C-10 column
- 19-5037-01Tube C 10/40, glass, inner Ø 10 mm