Orders LinkContactUsLinkComponent
Test tubes racks, wire
Test tubes racks, wire
Supplier:   HEATHROW
Test tubes racks, wire
Supplier:  HEATHROW
Certificates
Test tubes racks, wire
Test tubes racks, wire
2 Options Available Below
Product Details & Documents
Tube racks, made of chemically-resistant epoxy-coated steel wire.Durable racks for transport or sample handling.
  • With slotted bottom
  • Holds either 13 and 16 mm tubes
  • Very good chemical resistance, autoclavable
Product Family Options
Product Information
  • Farve
    Ant. huller
    Til rør Ø
    B×D×H
    Availability
    Price
  • rød
    108 (9×12)
    16 mm
    188×241×65 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 273,00 kr
    273,00 kr
    1 * 1 Items
       273,00 kr
  • blå
    72 (6×12)
    16 mm
    127×237×65 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 271,00 kr
    271,00 kr
    1 * 1 Items
       271,00 kr