Multi-way stopcocks, Discofix®
Leverandør: B. Braun
Gå til:
Om denne vare
Suitable for all pressure infusion systems <2 bar.
- Continuous flow channels for accurate pressure monitoring
- Swivel lock can be moved axially or radially for fast and secure connection
- PVC and latex-free
Suitable for all pressure infusion systems <2 bar.
8 tilgængelige muligheder nedenfor
Product Details & Documents
Suitable for all pressure infusion systems <2 bar.
- Continuous flow channels for accurate pressure monitoring
- Swivel lock can be moved axially or radially for fast and secure connection
- PVC and latex-free
Suitable for all pressure infusion systems <2 bar.
Documents
Product Family Options
Discofix®-2, two-way stopcock. Handle can be rotated through 180°
Produktinformation
Discofix®-3, three-way stopcock. Handle can be rotated through 360°
Produktinformation
- FarveTilgængelighedPrice
- 720-25743-vejs stophane
- BRAU4098218Three-way stopcocks with 250 mm extension
- BRAU4098080Three-way stopcocks with 1000 mm extension
- 720-25733-vejs stophane
- BRAU4098102Three-way stopcocks with 100 mm extension
- BRAU4098501Three-way stopcocks with 500 mm extension
- BRAU4095146Three-way stopcocks