Orders LinkContactUsLinkComponent
Buffer solutions in sachets, Certipur®, Supelco®
Buffer solutions in sachets, Certipur®, Supelco®
Leverandør:   Merck
Buffer solutions in sachets, Certipur®, Supelco®
Leverandør:  Merck
Certifikater
Buffer solutions in sachets, Certipur®, Supelco®
Buffer solutions in sachets, Certipur®, Supelco®
12 tilgængelige muligheder nedenfor
Product Details & Documents
Til kalibrering af pH-metre.
  • Practical 30 ml sachets - one use per sachet - no risk of contamination
  • Solutions are always fresh and ready to use
  • Traceable to SRM from NIST and PTB
  • Reference temperature: +25 °C
  • Precision: Deviation is max. ±0,015 pH units
pH value is measured with a combined glass electrode after 5-point calibration according to DIN 19268 with reference buffer solutions according to DIN 19266, IUPAC, NIST, Ph. Eur. and USP.
Specifications for Product Family
Specifikationer
Specifikationer
  • Catalog No:
  • 1.99012.0001
  • 1.99064.0001
  • 1.99001.0001
  • 1.99016.0001
  • 1.99002.0001
  • 1.99003.0001
  • 1.99019.0001
  • 1.99004.0001
  • 1.99021.0001
  • 1.99022.0001
  • 1.99005.0001
  • 1.99006.0001
  • M-Clarity quality level:
  • MQ100
  • MQ100
  • MQ100
  • MQ100
  • MQ100
  • MQ100
  • MQ100
  • MQ100
  • MQ100
  • MQ100
Specifikationer
Product Family Options
Produktinformation
  • pH-værdi
    pH buffer type
    Pakning
    Pk Info
    Pakn
    Tilgængelighed
    Price
  • 1.99012.0001
    Certipur® buffer solution, pH 2,00, citric acid, sodium hydroxide, hydrogen chloride
    Certipur® buffer solution, pH 2,00, citric acid, sodium hydroxide, hydrogen chloride
    2,00
    Citrat
    Sachet
    30×30 ml
    30
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 30 Items
    1 * 30 Items - 868,00 kr
    868,00 kr
    1 * 30 Items
       868,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    1.99012.0001
    M-Clarity quality level
    MQ100
  • 1.99064.0001
    Certipur® buffer solution, pH 4,00, citric acid, sodium hydroxide, hydrogen chloride
    Certipur® buffer solution, pH 4,00, citric acid, sodium hydroxide, hydrogen chloride
    4.00
    Citrat
    Sachet
    30×30 ml
    30
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 30 Items
    1 * 30 Items - 829,00 kr
    829,00 kr
    1 * 30 Items
       829,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    1.99064.0001
    M-Clarity quality level
  • 1.99001.0001
    CertiPUR® buffer solution, pH 4,01, potassium hydrogen phthalate
    CertiPUR® buffer solution, pH 4,01, potassium hydrogen phthalate
    4,01
    Phthalat
    Sachet
    30×30 ml
    30
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 30 Items
    1 * 30 Items - 868,00 kr
    868,00 kr
    1 * 30 Items
       868,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    1.99001.0001
    M-Clarity quality level
    MQ100
  • 1.99016.0001
    Certipur® buffer solution, pH 6,00, citric acid, sodium hydroxide
    Certipur® buffer solution, pH 6,00, citric acid, sodium hydroxide
    6,00
    Citrat
    Sachet
    30×30 ml
    30
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 30 Items
    1 * 30 Items - 829,00 kr
    829,00 kr
    1 * 30 Items
       829,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    1.99016.0001
    M-Clarity quality level
  • 1.99002.0001
    Certipur® buffer solution, pH 7,00, potassium hydrogen phosphate, disodium hydrogen phosphate
    Certipur® buffer solution, pH 7,00, potassium hydrogen phosphate, disodium hydrogen phosphate
    7,00
    Fosfat
    Sachet
    30×30 ml
    30
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 30 Items
    1 * 30 Items - 868,00 kr
    868,00 kr
    1 * 30 Items
       868,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    1.99002.0001
    M-Clarity quality level
    MQ100
  • 1.99003.0001
    Certipur® buffer solution, pH 9,00, boric acid, sodium hydroxide, potassium chloride
    Certipur® buffer solution, pH 9,00, boric acid, sodium hydroxide, potassium chloride
    9,00
    Borat
    Sachet
    30×30 ml
    30
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 30 Items
    1 * 30 Items - 829,00 kr
    829,00 kr
    1 * 30 Items
       829,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    1.99003.0001
    M-Clarity quality level
    MQ100
  • 1.99019.0001
    Certipur® buffer solution, pH 9,18, disodium tetraborate
    Certipur® buffer solution, pH 9,18, disodium tetraborate
    9.18
    Borat
    Sachet
    30×30 ml
    30
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 30 Items
    1 * 30 Items - 829,00 kr
    829,00 kr
    1 * 30 Items
       829,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    1.99019.0001
    M-Clarity quality level
    MQ100
  • 1.99004.0001
    CertiPur® buffer solution, pH 10,00, boric acid, sodium hydroxide, potassium chloride
    CertiPur® buffer solution, pH 10,00, boric acid, sodium hydroxide, potassium chloride
    10,00
    Borat
    Sachet
    30×30 ml
    30
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 30 Items
    1 * 30 Items - 829,00 kr
    829,00 kr
    1 * 30 Items
       829,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    1.99004.0001
    M-Clarity quality level
    MQ100
  • 1.99021.0001
    Certipur® buffer solution, pH 11,00, boric acid, sodium hydroxide, potassium chloride
    Certipur® buffer solution, pH 11,00, boric acid, sodium hydroxide, potassium chloride
    11.00
    Borat
    Sachet
    30×30 ml
    30
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 30 Items
    1 * 30 Items - 868,00 kr
    868,00 kr
    1 * 30 Items
       868,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    1.99021.0001
    M-Clarity quality level
    MQ100
  • 1.99022.0001
    Certipur® buffer solution, pH 12,00, potassium dihydrogen phosphate, sodium hydroxide
    Certipur® buffer solution, pH 12,00, potassium dihydrogen phosphate, sodium hydroxide
    12,00
    Fosfat
    Sachet
    30×30 ml
    30
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 30 Items
    1 * 30 Items - 829,00 kr
    829,00 kr
    1 * 30 Items
       829,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    1.99022.0001
    M-Clarity quality level
    MQ100
  • 1.99005.0001
    Certipur® buffer solution, set 1 (pH 4,01, 7,00 and 9,00)
    Certipur® buffer solution, set 1 (pH 4,01, 7,00 and 9,00)
    4,01/7,00/9,00
    Sachet
    30×30 ml
    30
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 30 Items
    1 * 30 Items - 829,00 kr
    829,00 kr
    1 * 30 Items
       829,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    1.99005.0001
    M-Clarity quality level
    MQ100
  • 1.99006.0001
    Certipur® buffer solution, set 2 (pH 4,01, 7,00 and 10,00)
    Certipur® buffer solution, set 2 (pH 4,01, 7,00 and 10,00)
    4,01/7,00/10,00
    Sachet
    30×30 ml
    30
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 30 Items
    1 * 30 Items - 868,00 kr
    868,00 kr
    1 * 30 Items
       868,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    1.99006.0001
    M-Clarity quality level
    MQ100