Liquid detergent, L900, Nalgene®
Liquid detergent, L900
Specifically formulated to clean plastic and glass labware used in pharmaceutical, medical research, academic, biotechnology and other facilities.
- Formulated to be compatible with all plastics, especially polycarbonate
- Will not cause crazing, stress cracking or clouding when used as directed
- For hand or machine use, it is non toxic and low foaming
Non inhibitory according to APHA Standard Methods for the Examination of Water and Wastewater.
- Beskrivelse:Liquid detergent, L900
- Cat. No.:148-0114
- Miljøcertificering:NAR Certificate of Retirement
- Environmentally Preferable:Lav fremstillingspåvirkning