Sample dippers, BOLA
Leverandør: Bohlender
Gå til:
Om denne vare
Sample vessels suitable for washing, rinsing or dipping solids in aggressive, or pure substances.
- Chemical resistance and temperature resistance
- Handle and holes in wall and botton
Certifications: FDA conform.
3 tilgængelige muligheder nedenfor
Product Details & Documents
Sample vessels suitable for washing, rinsing or dipping solids in aggressive, or pure substances.
- Chemical resistance and temperature resistance
- Handle and holes in wall and botton
Certifications: FDA conform.
Product Family Options
Produktinformation
- MaterialeLængdeØTilgængelighedPrice
- BOHLH1138-24Øsekar, med greb og hul (Ø 12 mm)
- BOHLH1138-16Øsekar, med greb og hul (Ø 8 mm)
- BOHLH1138-08Øsekar, med greb og hul (Ø 6 mm)