Orders LinkContactUsLinkComponent
XCHANGE® syringes
XCHANGE® syringes
XCHANGE® syringes
XCHANGE® syringes
XCHANGE® syringes
The XCHANGE® coupling system secures and locks the syringe in place. This is achieved by a coupling mechanical piece that captures the syringe and a magnet that automatically aligns – eliminating the need for springs, screws or alignment gauges. Once engaged the syringe is locked into position requiring zero holding force.
  • Injection of small and large volumes in one sample sequence
  • Adding of internal standards without cross-contamination
  • Preventative exchange of syringe when using harmful media
Product Family Options
XCHANGE® syringes for GC
  • Beskrivelse
    Måler
    Volumen
    Længde
    Udv.Ø
    Spidstype
  • XCHANGE® CTC PAL-XT syringe for GC (all metal plunger)
    23
    10 µl
    56 mm
    0,63 mm
    Cone
    1 * 1 Items
    1 * 1 Items - 954,00 kr
    954,00 kr
    1 * 1 Items
       954,00 kr
    Specifications
    Cat. No.
    548-1805
  • XCHANGE® CTC PAL-XT syringe for GC
    23
    10 µl
    56 mm
    0,63 mm
    Cone
    1 * 1 Items
    1 * 1 Items - 874,00 kr
    874,00 kr
    1 * 1 Items
       874,00 kr
    Specifications
    Cat. No.
    SGEA2928210
  • XCHANGE® CTC PAL-XT syringe for GC
    23
    25 µl
    56 mm
    0,63 mm
    Cone
    1 * 1 Items
    1 * 1 Items - 874,00 kr
    874,00 kr
    1 * 1 Items
       874,00 kr
    Specifications
    Cat. No.
    548-1807
  • XCHANGE® CTC PAL-XT syringe for GC
    23
    50 µl
    56 mm
    0,63 mm
    Cone
    1 * 1 Items
    1 * 1 Items - 874,00 kr
    874,00 kr
    1 * 1 Items
       874,00 kr
    Specifications
    Cat. No.
    548-1809
  • XCHANGE® CTC PAL-XT syringe for GC
    23
    100 µl
    56 mm
    0,63 mm
    Cone
    1 * 1 Items
    1 * 1 Items - 874,00 kr
    874,00 kr
    1 * 1 Items
       874,00 kr
    Specifications
    Cat. No.
    548-1811
  • XCHANGE® CTC PAL-XT syringe for GC
    23
    250 µl
    56 mm
    0,63 mm
    Cone
    1 * 1 Items
    1 * 1 Items - 874,00 kr
    874,00 kr
    1 * 1 Items
       874,00 kr
    Specifications
    Cat. No.
    548-1813
  • XCHANGE® CTC PAL-XT syringe for GC
    23
    1000 µl
    56 mm
    0,63 mm
    Bevelled
    1 * 1 Items
    1 * 1 Items - 928,00 kr
    928,00 kr
    1 * 1 Items
       928,00 kr
    Specifications
    Cat. No.
    548-1817
XCHANGE® syringes for LC
  • Beskrivelse
    Måler
    Volumen
    Længde
    Udv.Ø
    Spidstype
  • XCHANGE® CTC PAL-XT syringe for LC (all metal plunger)
    22
    10 µl
    56 mm
    0,71 mm
    LC
    1 * 1 Items
    1 * 1 Items - 874,00 kr
    874,00 kr
    1 * 1 Items
       874,00 kr
    Specifications
    Cat. No.
    548-1806
  • XCHANGE® CTC PAL-XT syringe for LC
    22
    25 µl
    56 mm
    0,71 mm
    LC
    1 * 1 Items
    1 * 1 Items - 874,00 kr
    874,00 kr
    1 * 1 Items
       874,00 kr
    Specifications
    Cat. No.
    548-1808
  • XCHANGE® CTC PAL-XT syringe for LC
    22
    50 µl
    56 mm
    0,71 mm
    LC
    1 * 1 Items
    1 * 1 Items - 874,00 kr
    874,00 kr
    1 * 1 Items
       874,00 kr
    Specifications
    Cat. No.
    548-1810
  • XCHANGE® CTC PAL-XT syringe for LC
    22
    100 µl
    56 mm
    0,71 mm
    LC
    1 * 1 Items
    1 * 1 Items - 874,00 kr
    874,00 kr
    1 * 1 Items
       874,00 kr
    Specifications
    Cat. No.
    548-1812
  • XCHANGE® CTC PAL-XT syringe for LC
    22
    250 µl
    56 mm
    0,71 mm
    LC
    1 * 1 Items
    1 * 1 Items - 874,00 kr
    874,00 kr
    1 * 1 Items
       874,00 kr
    Specifications
    Cat. No.
    548-1814
  • XCHANGE® CTC PAL-XT syringe for LC
    22
    500 µl
    56 mm
    0,71 mm
    LC
    1 * 1 Items
    1 * 1 Items - 928,00 kr
    928,00 kr
    1 * 1 Items
       928,00 kr
    Specifications
    Cat. No.
    548-1816
  • XCHANGE® CTC PAL-XT syringe for LC
    22
    1000 µl
    56 mm
    0,71 mm
    LC
    1 * 1 Items
    1 * 1 Items - 928,00 kr
    928,00 kr
    1 * 1 Items
       928,00 kr
    Specifications
    Cat. No.
    548-1818
Replacement needles for GC XCHANGE® syringes
  •  
    Længde
    Spidstype
    Udv.Ø
    Beskrivelse
    Måler
  • 56 mm
    Bevelled
    0,63 mm
    Replacement needles for XCHANGE® GC syringes, 1000 µl
    23
    1 * 5 Items
    1 * 5 Items - 496,00 kr
    496,00 kr
    1 * 5 Items
       496,00 kr
Replacement needles for LC XCHANGE® syringes
  •  
    Længde
    Spidstype
    Udv.Ø
    Beskrivelse
    Måler
  • 56 mm
    LC
    0,71 mm
    Replacement needles for XCHANGE® LC syringes, 1000 µl
    22
    1 * 5 Items
    1 * 5 Items - 496,00 kr
    496,00 kr
    1 * 5 Items
       496,00 kr