Syringes for HPLC autosamplers, Hitachi instruments
Katalog # 548-1655
Leverandør: Trajan Scientific and Medical
Specifikationer
- Til:L7200/7250, AS1000/2000/4000 LC
- Model:500C-HITACHI
- Volumen:500 µl
- Beskrivelse:Gas-tight, with M10 - 1,0 screw thread termination
- Pakn:1