Inserts, WHEATON®
0,25 ml, Glas, No, -
Katalog # WHEA225258
Leverandør: DWK Life Sciences
Gå til:
Product Details & Documents
Insert is made of durable polypropylene (PP) and is designed for vials with a step.
- Polypropylene construction ensures chemical resistance
- Making it suitable for a wide range of laboratory applications
This insert securely holds 0,25 ml of samples in place for precise measurements and efficient sample handling.
With this level insert, you can ensure accurate and reliable sample storage without the risk of contamination. Upgrade your laboratory workflow with this high-quality 0,25 ml Level Insert, designed to streamline your sample preparation process and enhance overall efficiency.
Specifikationer
- Kasket inkluderet:No
- Steril:-
- Kapacitet:0,25 ml
- Materiale:Glas