Orders LinkContactUsLinkComponent
Cell culture vessels, Corning® HYPERStack®
Leverandør:   Corning
Cell culture vessels, Corning® HYPERStack®
Leverandør:  Corning
Certifikater
Cell culture vessels, Corning® HYPERStack®
Cell culture vessels, Corning® HYPERStack®
4 tilgængelige muligheder nedenfor
Product Details & Documents
The Corning® HYPERStack® cell culture vessel is a closed system that allows fluid manipulations to occur through preassembled tubing sets. Two ³/₈" ID tubing ports for filling and venting allow filling via bag assembly, sterile bottle connection or pumping. The attached vent filters are 0,2 μm pore, hydrophobic membranes that allow air displacement during filling while minimising the risk of contamination.
  • Features 18,000 cm² cell growth area
  • Closed system approach with integrally incorporated USP
  • Class VI certified C-FLEX tubing for liquid handling manipulations
  • Low particulate assembly method acceptable for cell therapy applications
  • Corning proprietary gas-permeable film technology
  • Corning CellBIND surface treatment for optimal cell attachment and performance
  • USP Class VI virgin polystyrene provides excellent optical clarity and mechanical strength
  • Gamma sterilized to SAL 10⁻⁶ standard
  • 100% leak tested prior to shipping
Product Family Options
Produktinformation
  • Dyrkningsareal
    Lukkestørrelse
    Pakket
    Tilgængelighed
    Price
  • 734-3377
    HYPERStack® - 12 layer cell culture vessel, Corning® CellBIND® surface
    HYPERStack® - 12 layer cell culture vessel, Corning® CellBIND® surface
    6000 cm²
    uden cap
    1 per pack/4 per case
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 4 Items
    1 * 4 Items - 25.430,00 kr
    25.430,00 kr
    1 * 4 Items
       25.430,00 kr
  • 734-3378
    HYPERStack® - 12 layer cell culture vessel, non-treated surface
    HYPERStack® - 12 layer cell culture vessel, non-treated surface
    6000 cm²
    uden cap
    1 per pack/4 per case
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 4 Items
    1 * 4 Items - 26.630,00 kr
    26.630,00 kr
    1 * 4 Items
       26.630,00 kr
  • 734-3379
    HYPERStack® - 36 layer cell culture vessel, Corning® CellBIND® surface
    HYPERStack® - 36 layer cell culture vessel, Corning® CellBIND® surface
    6000 cm²
    uden cap
    1 per pack/2 per case
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 2 Items
    1 * 2 Items - 26.930,00 kr
    26.930,00 kr
    1 * 2 Items
       26.930,00 kr
  • 734-3380
    HYPERStack® - 36 layer cell culture vessel, non-treated surface
    HYPERStack® - 36 layer cell culture vessel, non-treated surface
    6000 cm²
    uden cap
    1 per pack/2 per case
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 2 Items
    1 * 2 Items - 26.140,00 kr
    26.140,00 kr
    1 * 2 Items
       26.140,00 kr
Accessories
Produktinformation
  • Katalog #
    Rør int.Ø
    Rør udv.Ø
    Tilgængelighed
    Price
  • Engangsslangesæt, ADCF C-flex, steril
    Engangsslangesæt, ADCF C-flex, steril
    734-2487
    9,5 mm
    12,7 mm
    Log ind for tilgængelighed
    1 * 2 Items - 1.520,00 kr
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Rulleflaske med Easy Grip-låg, 850 cm², PS, ubehandlet, steril
    Rulleflaske med Easy Grip-låg, 850 cm², PS, ubehandlet, steril
    734-2492
    Log ind for tilgængelighed
    1 * 40 Items - 5.600,00 kr
Produktinformation
  • Katalog #
    Rør int.Ø
    Rør udv.Ø
    Tilgængelighed
    Price
  • Engangsslangesæt, MPC, 0,2 μm filter, steril
    Engangsslangesæt, MPC, 0,2 μm filter, steril
    734-2488
    9,5 mm
    12,7 mm
    Log ind for tilgængelighed
    1 * 2 Items - 6.350,00 kr
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • HYPERStack-manipulator, rustfrit stål
    HYPERStack-manipulator, rustfrit stål
    734-2491
    Log ind for tilgængelighed
    1 * 1 Items - 207.220,00 kr