Dextran, M.W. ~ 500,000
Glass bottle for solids, 100 g
Katalog # SERA18695.03
Marketsource
Leverandør: Serva
Gå til:
Product Details & Documents
- CAS nummer:9004-54-0
- Formel:(C₆H₁₀O₅)n
- MDL nummer:MFCD00130935
- EINECS:232-677-5
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:100 g
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



