Orders LinkContactUsLinkComponent
Cell counters
Cell counters
Leverandør:   HEATHROW
Cell counters
Leverandør:  HEATHROW
Certifikater
Cell counters
2 tilgængelige muligheder nedenfor
Product Details & Documents
These laboratory cell counters can measure cells such as basophils, eosinophils, myelocytes, juvenile, etc.
  • Each key records up to 999
  • Alarm at one hundred totals
  • Red and white buttons with grey housing
They can measure the number of cells such as Basophils (BASO), Eosinophils (EOSIN), Myelocytes (MYELO), Juvenile (JUVEN), Stab., Seg., Lymph., Mono., and total.
Product Family Options
Produktinformation
  • Matrix
    Tilgængelighed
    Price
  • HEATHS5360A
    Benchtop counter, 250×76×56 mm
    Benchtop counter, 250×76×56 mm
    6
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 3.650,00 kr
    3.650,00 kr
    1 * 1 Items
       3.650,00 kr
  • HEATHS5360B
    Benchtop counter, 320×76×56 mm
    Benchtop counter, 320×76×56 mm
    9
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 5.250,00 kr
    5.250,00 kr
    1 * 1 Items
       5.250,00 kr