pH-indikatorpapir, ruller, Whatman™
WHAT10362030
- pH-område:1,0 - 11,0
- Beskrivelse:Universalindikator, rulle
- Pakn:1 Roll
Universal indicator papers which have been impregnated with a mixture of several indicators. On contact with the sample solution they assume a particular colour. A check against the colour comparision table supplied allows the pH to be determined.
- Instant pH readings
- Accurate for a wide range of routine pH testing
- Convenient and portable for field use