Disposable earplugs, MultiMax®
Earplugs without cord, refill for dispenser LS-500, orange/gul, U*
Katalog # DALL3301261
Leverandør: HONEYWELL SAFETY
Gå til:
Product Details & Documents
These earplugs made of self-adjusting foam feature a two-sizes-in-one design and therefore provide optimal individual fit.
- Smooth, soil-resistant closed cell-foam skin prevents dirt build-up
- Self-adjusting polyurethane foam expands to fit virtually every wearer
- Save time, waste and space
SNR: 35 dB
: EN 352-2
Specifikationer
- Farve:orange/gul
- Beskrivelse:Earplugs without cord, refill for dispenser LS-500
- Seq:03
- Størrelse:U*
Accessories
Produktinformation
- Katalog #FarveTilgængelighedPrice
- Ørepropdispenser LS-500, tom, vægmontering, 500 par uden snorØrepropdispenser LS-500, tom, vægmontering, 500 par uden snorDALL3301273blåLog ind for tilgængelighed1 * 1 Items - 3.380,00 kr