Orders LinkContactUsLinkComponent
Reflectoquant® teststrimler, Supelco®
Reflectoquant® teststrimler, Supelco®
Blank strip uden reagens (til farvekorrektion), 50 Tests
Catalog # 1.16730.0001
Supplier:   Merck
Reflectoquant® teststrimler, Supelco®
Catalog # 1.16730.0001
Supplier:  Merck
CAS Number:  
Reflectoquant® teststrimler, Supelco®
Blank strip uden reagens (til farvekorrektion), 50 Tests
CAS Number:  
Price
336,00 kr
Log in for availability
1 * 1 Each
Product Details & Documents
Test strips for the reflectometers RQflex® 10, RQflex® 10 plus and RQflex® 20.
  • Fast analysis with reliable results
  • Barcode calibration
  • Low analysis costs
  • Environmentally friendly
Documents
Specifications
Specifications
  • Pakn:
    50 Tests
  • Beskrivelse:
    Blank strip uden reagens (til farvekorrektion)
Specifications
Complementary Products
Product Information
  • Catalog #
    Beskrivelse
    Model
    Availability
    Price
  • 1.17246.0001
    Reflectometer for test strips only
    RQflex 20
    Log in for availability
    1 * 1 Items - 10.580,00 kr