Reflectoquant® teststrimler, Supelco®
Test strips for the reflectometers RQflex® 10, RQflex® 10 plus and RQflex® 20.
- Fast analysis with reliable results
- Barcode calibration
- Low analysis costs
- Environmentally friendly
Nitrite is naturally present in soil, water and food, and regularly testing its concentration is essential for many applications. Nitrites must be closely monitored in drinking and groundwater as well as in process, waste, or aquarium water. In farm produce and fruit juices, high nitrate concentrations affect quality and keeping properties and can constitute serious health risks. Nitrites are also used in additives for food products and serve as corrosion inhibitors in engine cooling systems.
Test strips for the reflectometers RQflex® 10, RQflex® 10 plus and RQflex® 20.
- Fast analysis with reliable results
- Barcode calibration
- Low analysis costs
- Environmentally friendly
Nitrite is naturally present in soil, water and food, and regularly testing its concentration is essential for many applications. Nitrites must be closely monitored in drinking and groundwater as well as in process, waste, or aquarium water. In farm produce and fruit juices, high nitrate concentrations affect quality and keeping properties and can constitute serious health risks. Nitrites are also used in additives for food products and serve as corrosion inhibitors in engine cooling systems.
- Catalog No:
- 1.16973.0001
- 1.16732.0001
- M-Clarity quality level:
- MQ100
- MQ100
Product Family Options
- BeskrivelseOmrådeKemisk metodePakn
- Nitrittest0,5 - 25,0 mg/l NO₂Griess' reaction50 Tests1 * 1 Each422,00 kr
Specifications
Cat. No.1.16973.0001M-Clarity quality levelMQ100 - Nitrattest30 - 1000 mg/l NO₂Aromatic amine50 Tests1 * 1 Each844,00 kr
Specifications
Cat. No.1.16732.0001M-Clarity quality levelMQ100