Reflectoquant® teststrimler, Supelco®
Glucosetest, 1,0 - 100 mg/l C₆H₁₂O₆, Enzymatic reaction, 50 Tests
: Merck
Test strips for the reflectometers RQflex® 10, RQflex® 10 plus and RQflex® 20.
- Fast analysis with reliable results
- Barcode calibration
- Low analysis costs
- Environmentally friendly
- Kemisk metode:Enzymatic reaction
- Pakn:50 Tests
- Beskrivelse:Glucosetest
- Område:1,0 - 100 mg/l C₆H₁₂O₆
- Cat. No.:1.16720.0001
- M-Clarity quality level:MQ100