Reflectoquant® teststrimler, Supelco®
Jerntest, 0,5 - 20,0 mg/l Fe(II), Triazine, 50 Tests
: Merck
Test strips for the reflectometers RQflex® 10, RQflex® 10 plus and RQflex® 20.
- Fast analysis with reliable results
- Barcode calibration
- Low analysis costs
- Environmentally friendly
- Kemisk metode:Triazine
- Pakn:50 Tests
- Beskrivelse:Jerntest
- Område:0,5 - 20,0 mg/l Fe(II)
- Cat. No.:1.16982.0001
- M-Clarity quality level:MQ100