Product Details & Documents
Test strips for the reflectometers RQflex® 10, RQflex® 10 plus and RQflex® 20.
- Fast analysis with reliable results
- Barcode calibration
- Low analysis costs
- Environmentally friendly
Documents
msds_116896
SDS
Specifikationer
Specifikationer
- Kemisk metode:Redox reaction
- Pakn:50 Tests
- Beskrivelse:Chlortest med væskereagens (frit chlor)
- Model:RQ
- Område:0,5 - 10,0 mg/l Cl₂
- Cat. No.:1.16896.0001
- M-Clarity quality level:MQ100
Specifikationer
Complementary Products
Produktinformation
- Katalog #BeskrivelseModelTilgængelighedPrice
- 1.17246.0001Reflectometer for test strips onlyRQflex 20Log ind for tilgængelighed1 * 1 Items - 10.580,00 kr