Product Details & Documents
Test strips for the reflectometers RQflex® 10, RQflex® 10 plus and RQflex® 20.
- Fast analysis with reliable results
- Barcode calibration
- Low analysis costs
- Environmentally friendly
Documents
Specifikationer
Specifikationer
- Kemisk metode:Phthalein complexone
- Pakn:50 Tests
- Beskrivelse:Calciumtest
- Område:5 - 125 mg/l Ca
- Cat. No.:1.16125.0001
- M-Clarity quality level:MQ100
Specifikationer
Complementary Products
Produktinformation
- Katalog #BeskrivelseModelTilgængelighedPrice
- 1.17246.0001Reflectometer for test strips onlyRQflex 20Log ind for tilgængelighed1 * 1 Items - 10.580,00 kr