Orders LinkContactUsLinkComponent
HPLC distributors for bottles
GL 45, 4 × max. 1,6 - 4 mm
Catalog # BOHLD606-08
Supplier:   Bohlender
HPLC distributors for bottles
Catalog # BOHLD606-08
Supplier:  Bohlender
HPLC distributors for bottles
HPLC distributors for bottles
GL 45, 4 × max. 1,6 - 4 mm
Price
236,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Green screw cap, made of glass-fibre reinforced PP, and distributor body made of PP.
  • Very good chemical resistance
  • Temperature resistant from −50 to +100 °C
Suitable for GL 45 threaded bottles and it is available with the plugs for closing unused ports.
Specifications
  • Gevind:
    GL 45
  • Rør udv. Ø:
    4 × max. 1,6 - 4 mm