Orders LinkContactUsLinkComponent
Sølvnitrat
Sølvnitrat
Oxidizing
Corrosive
Environment
Marketsource
Supplier:   Roth Carl
Sølvnitrat
Supplier:  Roth Carl
Marketsource
Certificates
Sølvnitrat
3 Options Available Below
Product Details & Documents
Synonyms:  Lunar caustic, Silver mononitrate, Nitric acid silver salt
  • CAS Number:7761-88-8
  • Formula:AgNO₃
  • MDL Number:MFCD00003414
  • Molecular Weight:169.87 g/mol
  • Merck Index:12,08661
  • Boiling Pt:444 °C (1013 hPa)
  • Melting Pt:210 °C
  • Density:4,35 g/cm³ (20 °C)
  • Storage Temperature:Stue temperatur
  • EINECS:231-853-9
  • UN:1493
  • ADR:5.1,II
  • REACH:01-2119513705-43
Product Family Options
Product Information
  • Pakn
    Availability
    Price
  • 50 g
    Log in for availability
    Log in for availability
    1 * 50 g
    1 * 50 g
    Specifications:
    Marketsource
    Marketsource
  • 250 g
    Log in for availability
    Log in for availability
    1 * 250 g
    1 * 250 g
    Specifications:
    Marketsource
    Marketsource
  • 1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg
    Specifications:
    Marketsource