(E)-Crotonaldehyd
Glass bottle, 250 mL
Katalog # ROTH3425.1
Marketsource
Leverandør: Roth Carl
Gå til:
Product Details & Documents
Synonymer: Crotonaldehyde
- CAS nummer:123-73-9
- Formel:C₄H₆O
- MDL nummer:MFCD00007003
- Molecular Weight:70.09 g/mol
- EINECS:204-647-1
Specifikationer
Specifikationer
- Pakning:Glass bottle
- Pakn:250 mL



