Phenylacetamid
Glass bottle for solids, 100 mg
Catalog # PROOMM0169.02
Supplier: LGC Standards PROMOCHEM
Jump to:
Product Details & Documents
Synonyms: 2-Phenylacetamide
- CAS Number:103-81-1
- Formula:C₈H₉NO
- MDL Number:MFCD00059193
- Molecular Weight:135.17 g/mol
- EINECS:203-147-0
Specifications
Specifications
- Pakning:Glass bottle for solids
- Pakn:100 mg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



