Pasteur pipettes, disposable
Leverandør: MARIENFELD
Gå til:
Om denne vare
Pasteur pipettes made of soda-lime glass.
- For single use
- Long, fine tip
- Constricted upper suction end to retain a cotton plug (supplied without cotton plug)
- Handy packaging in boxes containing 250 pieces
Certifications: In compliance with DIN ISO 7712
2 tilgængelige muligheder nedenfor
Product Details & Documents
Pasteur pipettes made of soda-lime glass.
- For single use
- Long, fine tip
- Constricted upper suction end to retain a cotton plug (supplied without cotton plug)
- Handy packaging in boxes containing 250 pieces
Certifications: In compliance with DIN ISO 7712
Product Family Options
Produktinformation
- Udv.ØLængdePakketTilgængelighedPrice
- MARI3233049Pasteur pipette, unpluggedPasteur pipette, unplugged1,3 - 1,4 mm150 mm4×250 piecesLog ind for tilgængelighed1 * 1.000 Itemskr836.00
- MARI3233050Pasteurpipette, utilproppetPasteurpipette, utilproppet1,3 - 1,4 mm230 mm4×250 stk.Log ind for tilgængelighed1 * 1.000 Itemskr852.00